Learn More
Description
HA726-305 is for Hitachi 704. HA926-610 is for Hitachi 911, 912, 914. HA426-R1 and HA426-R2 for Hitachi 747.
- Method: Modified IFCC, Kinetic
- Format: Liquid
- Linearity: 0-500 U/L
- Expected Values: 4-36 U/L
- Storage Temperature: 2° to 8°C
- Reconstituted Stability: Ready to use liquid
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | ALT R1 Kit |
| Formulation | Ketoglutaric Acid 0.2%, Sodium Azide 0.095% |
| For Use With (Application) | Quantitative determination of Alanine Aminotransferase in Serum |
| For Use With (Equipment) | Hitachi 747 analyzer |
| Quantity | 5 x 500 R1mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.