Learn More
Description
Reagent Set 1 includes 1 x 120mL substrate/enzyme reagent and 1 x 30mL coenzyme reagent.
Reagent Set 2 includes 3 x 120mL substrate/enzyme reagent and 1 x 90mL coenzyme reagent.
Reagent Set 3 includes 1 x 500mL substrate/enzyme reagent and 1 x 125mL coenzyme reagent.
- Method: Modified IFCC, Kinetic
- Format: Liquid
- Wavelength: 340nm
- Linearity: 0-500 U/L
- Expected Values: 4-36 U/L
- Storage Temperature: 2° to 8°C
- Reconstituted Stability: Ready to use liquid
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Alanine Aminotransferase Reagent |
| Formulation | L-alanine 500mM, alpha-Ketoglutaric Acid 15mM, LDH(microbial) >2000IU/L, NADH >0.18mM, Buffer 100mM pH 7.5 ±0.1, Sodium Azide 0.25%, Stabilizers |
| For Use With (Application) | Quantitative determination of Alanine Aminotransferase in Serum |
| Quantity | 3 x 120mL R1, 1 x 90mL R2 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.