Learn More
Description
HA965-324 is for Hitachi 911, 912, 914. HA765-162 is for Hitachi 704. HA465-R1 and HA465-R2 are for Hitachi 747.
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Amylase-EPS R1 |
| Formulation | Sodium Azide 0.095% |
| For Use With (Application) | Quantitative kinetic determination of a-amylase activity in Human Serum |
| For Use With (Equipment) | Hitachi 747 analyzers |
| Quantity | 3 x 270 R1mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.