Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Amylase Reagents

Catalog No. 23666111 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
2 x 30 mL
4 x 30 mL
4 x 120 mL
1 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-111 2 x 30 mL
23-666-113 4 x 30 mL
23-666-112 4 x 120 mL
23-666-110 1 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-666-111 Supplier Pointe Scientific Supplier No. A756460
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

MES Buffer pH 6.0±0.1; 2-Chloro-p-Nitrophenyl-α-D-Maltotrioside1.8mM; Sodium Chloride 350mM; Calcium Acetate 6mM; Potassium Thiocyanate 900mM; Sodium Azide 0.1%

Test α-amylase activity for diagnoses

  • Method: Kinetic, CNP3G
  • Format: Liquid (ready to use)
  • Wavelength: 405nm
  • Linearity: 0 to 2000U/L
  • Expected Values: 25 to 125U/L

For quantitative kinetic determination of α-amylase activity in human serum. Most commonly used for diagnosis and treatment of pancreatic diseases.

Specifications

Certifications/Compliance CE Marked
Type Amylase Reagent
Formulation MES Buffer pH 6, 2-Chloro-p-Nitrophenyl-alpha-D-Maltotrioside 1.8mM, Sodium Chloride 350mM, Calcium Acetate 6mM, Potassium Thiocyanate 900mM, Sodium Azide 0.1%
For Use With (Application) Quantitative kinetic determination of a-amylase activity in Human Serum
Quantity 2 x 30 mL
Wavelength 405 nm
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.