Learn More
Description
Includes
MES Buffer pH 6.0±0.1; 2-Chloro-p-Nitrophenyl-α-D-Maltotrioside1.8mM; Sodium Chloride 350mM; Calcium Acetate 6mM; Potassium Thiocyanate 900mM; Sodium Azide 0.1%
- Method: Kinetic, CNP3G
- Format: Liquid (ready to use)
- Wavelength: 405nm
- Linearity: 0 to 2000U/L
- Expected Values: 25 to 125U/L
For quantitative kinetic determination of α-amylase activity in human serum. Most commonly used for diagnosis and treatment of pancreatic diseases.
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Amylase Reagent |
| Formulation | MES Buffer pH 6, 2-Chloro-p-Nitrophenyl-alpha-D-Maltotrioside 1.8mM, Sodium Chloride 350mM, Calcium Acetate 6mM, Potassium Thiocyanate 900mM, Sodium Azide 0.1% |
| For Use With (Application) | Quantitative kinetic determination of a-amylase activity in Human Serum |
| Quantity | 4 x 120 mL |
| Wavelength | 405 nm |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.