Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific AST Reagents

Catalog No. 23666128 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
5 x 253 mL
5 x 50 mL R1, 1 x 50 mL R2
5 x 500 mL
Type:
AST R1
AST R2
AST Reagent Kit
For Use With (Equipment):
Hitachi 704 Analyzers
Hitachi 747 Analyzers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type For Use With (Equipment)
23-666-128 5 x 50 mL R1, 1 x 50 mL R2 AST Reagent Kit Hitachi 704 Analyzers
23-666-131 5 x 500 mL AST R1 Hitachi 747 Analyzers
23-666-132 5 x 253 mL AST R2 Hitachi 747 Analyzers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-128 Supplier Pointe Scientific Supplier No. HA761305

For quantitative determination of Aspartate Aminotransferase (AST) in human serum on Hitachi analyzers

HA761-305 is for Hitachi 704. HA961-610 is for Hitachi 911, 912, 914. HA461-R1 and HA461-R2 are for Hitachi 747.

  • Modified IFCC, Kinetic
  • Format: Liquid
  • Linearity: 0-500 U/L
  • Expected Values: 5-34 U/L
  • Reconstituted Stability: Ready to use liquid

Specifications

Certifications/Compliance CE Marked
Type AST Reagent Kit
Formulation L-aspartic Acid >200mM, alpha-Ketoglutaric Acid 12mM, LDH (microbial) >1000U/L, MDH (microbial) >800U/L, NADH >0.18mM, pH 7.8 ±0.1 Buffer, Sodium Azide 0.25%, Stabilizers
For Use With (Application) Quantitative determination of Aspartate Aminotransferase in Human Serum
For Use With (Equipment) Hitachi 704 Analyzers
Quantity 5 x 50 mL R1, 1 x 50 mL R2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.