Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific AST (SGOT) Liquid Reagents

Catalog No. 23666120 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120 mL R1, 1 x 30 mL R2
1 x 500 mL R1, 1 x 125 mL R2
3 x 120 mL R1, 1 x 90 mL R2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-120 1 x 120 mL R1, 1 x 30 mL R2
23-666-121 3 x 120 mL R1, 1 x 90 mL R2
23-666-122 1 x 500 mL R1, 1 x 125 mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-120 Supplier Pointe Scientific Supplier No. A7561150
Add to Cart
Add to Cart
This item is not returnable. View return policy

For quantitative determination of Aspartate Aminotransferase (AST) in human serum

Set 1 consists of 1 x 120mL substrate reagent and 1 x 30mL enzyme reagent.
Set 2 consists of 3 x 120mL substrate reagent and 1 x 90mL enzyme reagent.
Set 3 consists of 1 x 500mL substrate reagent and 1 x 125mL enzyme reagent.

  • Method: Modified IFCC, Kinetic
  • Format: Liquid
  • Linearity: 0-500 U/L
  • Expected Values: 4-36 U/L
  • Reconstituted Stability: Ready to use liquid

Specifications

Certifications/Compliance CE Marked
Type Aspartate Aminotransferase Reagent Set 1
Formulation L-aspartic Acid >200mM, alpha-Ketoglutaric Acid 12mM, LDH (microbial) >1000U/L, MDH (microbial) >800U/L, NADH >0.18mM, pH 7.8 ±0.1 Buffer, Sodium Azide 0.25%, Stabilizers
For Use With (Application) Quantitative determination of Aspartate Aminotransferase in Human Serum
Quantity 1 x 120 mL R1, 1 x 30 mL R2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.