Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific autoHDL Cholesterol Liquid Reagents

Catalog No. 23666137 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 240L R1, 1 x 800mL R2
1 x 30mL R1, 1 x 10mL R2
1 x 60mL R1, 1 x 20mL R2
1 x 750mL R1, 1 x 250mL R2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-137 1 x 240L R1, 1 x 800mL R2
23-666-136 1 x 750mL R1, 1 x 250mL R2
23-666-138 1 x 30mL R1, 1 x 10mL R2
23-666-139 1 x 60mL R1, 1 x 20mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-666-137 Supplier Pointe Scientific Supplier No. H7545320
Add to Cart
Add to Cart
This item is not returnable. View return policy

For quantitative determination high-density lipoprotein cholesterol in human serum or plasma

Reagent requires autoHDL/LDL Calibrator (23-666-140).

  • Method: Direct/Automated
  • Format: Liquid
  • Wavelength: 600/70nm
  • Linearity: 2 - 150mg/dL
  • Expected Values: 30-85mg/dL
  • Storage Temperature: 2° to 8°C
  • Reconstituted Stability: Ready to use liquid

Specifications

Certifications/Compliance CE Marked
Type AutoHDL™ Cholesterol Liquid Reagent Kit
For Use With (Application) Quantitative determination high-density Lipoprotein Cholesterol in Human Serum or Plasma
Quantity 1 x 240L R1, 1 x 800mL R2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.