Learn More
Description
Reagent requires autoHDL/LDL Calibrator (23-666-140).
- Method: Direct/Automated
- Format: Liquid
- Wavelength: 600/70nm
- Linearity: 2 - 150mg/dL
- Expected Values: 30-85mg/dL
- Storage Temperature: 2° to 8°C
- Reconstituted Stability: Ready to use liquid
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | AutoHDL™ Cholesterol Liquid Reagent Kit |
| For Use With (Application) | Quantitative determination high-density Lipoprotein Cholesterol in Human Serum or Plasma |
| Quantity | 1 x 30mL R1, 1 x 10mL R2 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.