Learn More
Description
Requires autoHDL/LDL Calibrator (23-666-140). HH945-240 is for Hitachi 911, 912, 914. HH445-R1 and HH4450R2 are for Hitachi 747.
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | AutoHDL Cholesterol Reagent Kit |
| For Use With (Application) | Quantitative determination high-density Lipoprotein Cholesterol in Human Serum or Plasma |
| Quantity | 6 x 30mL R1, 3 x 20mL R2 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.