Learn More
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Bilirubin Reagent Kit |
| Formulation | Direct Bilirubin reagent includes 32mM Sulfanilic Acid in dilute Hydrochloric Acid, Sodium Nitrite reagent includes Sodium Nitrite 60mM |
| For Use With (Application) | Quantitative determination of direct Bilirubin in Serum, in-vitro diagnostic use only |
| pH | 1.1 |
| Quantity | 1L Reagent, 15mL Nitrite |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.