Learn More
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Bilirubin Reagent Kit |
| Formulation | Direct Bilirubin R1 includes Acid Buffer 50mM/L, direct Bilirubin R2 includes Acid Buffer >30mM/L, DPD and Stabilizers >2mM/L |
| For Use With (Application) | Quantitative determination of direct Bilirubin in Serum, in-vitro diagnostic use only |
| pH | 1.7 |
| Quantity | 4 x 300 R1mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.