Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Bilirubin Direct Liquid Reagents

Catalog No. 23666464 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
120mL Reagent, 15mL Nitrite
1L Reagent, 15mL Nitrite
4 x 100 R2mL
4 x 300 R1mL
500mL Reagent, 15mL Nitrite
6 x 19mL R1, 3 x 11mL R2
Type:
Bilirubin Reagent Kit
Bilirubin Reagent Set
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-666-464 4 x 100 R2mL Bilirubin Reagent Kit
23-666-147 120mL Reagent, 15mL Nitrite Bilirubin Reagent Kit
23-666-148 1L Reagent, 15mL Nitrite Bilirubin Reagent Kit
23-666-149 500mL Reagent, 15mL Nitrite Bilirubin Reagent Kit
23-666-463 4 x 300 R1mL Bilirubin Reagent Kit
23-666-466 6 x 19mL R1, 3 x 11mL R2 Bilirubin Reagent Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 23-666-464 Supplier Pointe Scientific Supplier No. HB436R2

Specifications

Certifications/Compliance CE Marked
Type Bilirubin Reagent Kit
Formulation Direct Bilirubin R1 includes Acid Buffer 50mM/L, direct Bilirubin R2 includes Acid Buffer >30mM/L, DPD and Stabilizers >2mM/L
For Use With (Application) Quantitative determination of direct Bilirubin in Serum, in-vitro diagnostic use only
pH 1.7
Quantity 4 x 100 R2mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.