Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Bilirubin Total Reagents for Hitachi Analyzers

Catalog No. 23666452 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
5 x 175 R2mL
5 x 600 R1mL
6 x 45mL R1, 3 x 25mL R2
Formulation:
Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
R1 contains: Acid Buffer 50mM/L, Surfactant, R2 contains: Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
Total Bilirubin R1 Acid Buffer 50mM/L, Surfactants, Total Bilirubin R2 Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
23-666-452 5 x 175 R2mL Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
23-666-465 5 x 600 R1mL Total Bilirubin R1 Acid Buffer 50mM/L, Surfactants, Total Bilirubin R2 Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
23-666-469 6 x 45mL R1, 3 x 25mL R2 R1 contains: Acid Buffer 50mM/L, Surfactant, R2 contains: Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-666-452 Supplier Pointe Scientific Supplier No. HB479R2

Specifications

Certifications/Compliance CE Marked
Type Bilirubin Total Reagent
Formulation Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
For Use With (Application) Quantitative determination of total Bilirubin in Serum
Quantity 5 x 175 R2mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.