Learn More
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | BUN Reagent Kit |
| Formulation | Urease (Jack Bean) >15000U/L, GLDH (Bovine) >200U/L, ADP >0.6mM, alpha-Ketoglutarate 3.6mM, NADH >0.28mM, Buffer pH 7.8 ± 0.1, Stabilizers, Sodium Azide 0.25% as preservative |
| For Use With (Application) | Quantitative determination of Urea Nitrogen in Serum |
| For Use With (Equipment) | Hitachi 704 analyzers |
| Quantity | 8 x 50mL R1, 2 x 50mL R2 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.