Learn More
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | BUN R1 |
| Formulation | Ketoglutaric Acid 0.2%, Sodium Azide 0.095% |
| For Use With (Application) | Quantitative determination of Urea Nitrogen in Serum |
| For Use With (Equipment) | Hitachi 747 analyzers |
| Quantity | 5 x 600 R1mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.