Learn More
Description
Includes
Calcium reagent: Arsenazo III≥0.15mM, 8-Hydroxyquinoline Sulfonate 5.0mM, buffer, surfactant
Requires
Requires Calibrator/Standard
- Method: Arsenazo III, Endpoint
- Format: Liquid (ready to use)
- Wavelength: 650nm
- Linearity: 0 to 15mg/dL
- Expected Values: 8.5 to 10.4mg/dL
For the quantitative determination of Calcium in serum or heparinized plasma.
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Calcium Arsenazo III Reagent Kit |
| Formulation | Calcium reagent: Arsenazo III <0.15mM, 8-Hydroxyquinoline Sulfonate 5mM, Buffer, Surfactant |
| For Use With (Application) | Quantitative determination of Calcium in Serum or Heparinized Plasma |
| Quantity | 1 x 1 L |
| Wavelength | 650 nm |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.