Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific CK (Liquid) Reagents

Catalog No. 23666209 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
150 mL
450 mL
625 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-209 450 mL
23-666-208 150 mL
23-666-210 625 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-209 Supplier Pointe Scientific Supplier No. C7522450
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Type Creatine Kinase Reagent Kit
Formulation CK R1 (Buffer) contains: Imidazole Buffer 100mM/L (pH 6.7), NADP 2mM/L, HK (Baker's Yeast) 2.5KU/L, Glucose 20mM/L, Magnesium Acetate 10mM/L, EDTA 2mM/L, N-acetylcysteine (NAC) 20mM/L, CK R2 (enzyme reagent) contains: Imidazole Buffer 100mM/L (pH 6.7), A
For Use With (Application) Quantitative determination of Creatine kinase activity in Serum
Quantity 450 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.