Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Creatinine (Cold Stable) Reagents

Catalog No. 23666237 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120mL R1, 1 x 30mL R2
1 x 1L R1, 1 x 250mL R2
1 x 500mL R1, 1 x 125mL R2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-237 1 x 120mL R1, 1 x 30mL R2
23-666-236 1 x 1L R1, 1 x 250mL R2
23-666-238 1 x 500mL R1, 1 x 125mL R2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-237 Supplier Pointe Scientific Supplier No. C7539150
Add to Cart
Add to Cart
This item is not returnable. View return policy

  • Reagent requires Creatinine Standard (23-666-235)/Chemistry Calibrator (23-666-189)

Specifications

Certifications/Compliance CE Marked
Type Creatinine Reagent Set
Formulation Creatinine R1: Alkaline Buffer, Creatinine R2: Picric Acid 40mM, Surfactant
For Use With (Application) Quantitative determination of Creatinine in Serum, in-vitro diagnostic use only
Quantity 1 x 120mL R1, 1 x 30mL R2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.