Learn More
Description
- Method: immunoturbidimetric
- Latex particle enhanced assay
- Reconstituted stability: ready-to-use liquid
- Linearity: 0.1 to 320.0mg/L
- Wavelength: 570nm
- Expected values: <3mg/L
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | CRP (hs) Wide Range Reagent Set |
| Formulation | R1: Buffer reagent Glycine Buffer 170mM, R2: Latex Suspension 0.20% (w/v), Latex particles coated with Rabbit Anti-Human CRP antibodies |
| For Use With (Application) | Quantitative determination of C-reactive Protein in Serum or Plasma |
| Quantity | 1 x 20 mL R1, 1 x 20 mL R2 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.