Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific CRP (hs) Wide Range Reagent Sets

Catalog No. 23666460 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 20 mL R1, 1 x 20 mL R2
1 x 50 mL R1, 1 x 50 mL R2
5 x 2 mL
6 x 3 mL
Type:
CRP (hs) Calibrator Set
CRP (hs) Control Set
CRP (hs) Wide Range Reagent Set
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-666-460 1 x 50 mL R1, 1 x 50 mL R2 CRP (hs) Wide Range Reagent Set
23-666-461 5 x 2 mL CRP (hs) Calibrator Set
23-666-462 6 x 3 mL CRP (hs) Control Set
23-666-459 1 x 20 mL R1, 1 x 20 mL R2 CRP (hs) Wide Range Reagent Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-666-460 Supplier Pointe Scientific Supplier No. C7568100
Add to Cart
Add to Cart
This item is not returnable. View return policy

For quantitative determination of C-reactive protein in serum or plasma

  • Method: immunoturbidimetric
  • Latex particle enhanced assay
  • Reconstituted stability: ready-to-use liquid
  • Linearity: 0.1 to 320.0mg/L
  • Wavelength: 570nm
  • Expected values: <3mg/L

Specifications

Certifications/Compliance CE Marked
Type CRP (hs) Wide Range Reagent Set
Formulation R1: Buffer reagent Glycine Buffer 170mM, R2: Latex Suspension 0.20% (w/v), Latex particles coated with Rabbit Anti-Human CRP antibodies
For Use With (Application) Quantitative determination of C-reactive Protein in Serum or Plasma
Quantity 1 x 50 mL R1, 1 x 50 mL R2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.