Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific General Chemistry Controls
Click to view available options
Type:
Ammonia or Alcohol Control Set
Chemistry Control (Level I)
Chemistry Control (Level II)
Lipid Control Set (Levels I and II)
Quantity:
2 x 5 mL
6 x 3 mL
10 x 5 mL
Description
- Human Serum Based (Lyophilized): Chemistry Control (Level 1)(23-666-187), Chemistry Control (Level II)(23-666-188), and Chemistry Control (Level 1 and II)(NC9896982)
- Hemoglobin Control Set (23-666-307) contains Human Erythrocytes (Lyophilized). Contains three levels of hemoglobin.
- Lipid Control Set (23-666-359) is Human Serum Based (Lyophilized). Expected Values: The lipid values were determined by repetitive assays of the listed methods. Values listed are targets only.
- Measurements using other reagents and instrument systems may give different results
- Listed ranges obtained from those suggested by the Health Care Finance Administration as acceptable performance criteria for deviation from a target value
- Due to method differences and matrix interactions, the values listed may not always be reproduced by other analytical systems
- Each laboratory should establish its own precision parameters
Specifications
Specifications
Certifications/Compliance | CE |
Content And Storage | 2°C to 8°C |
Type | Chemistry Control (Level II) |
Intended Use | In-vitro diagnostic use, monitoring the accuracy and precision of clinical chemistry procedures |
Level | 2 |
Quantity | 10 x 5 mL |
Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction