Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific General Chemistry Reagents

Catalog No. 23666080 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
150 mL
2 x 150 R2mL
450 mL
5 x 500 R1mL
625 mL
Type:
Alkaline Phosphatase R1 Kit
Alkaline Phosphatase R2 Kit
Alkaline Phosphatase Reagent Kit
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-666-080 5 x 500 R1mL Alkaline Phosphatase R1 Kit
23-666-074 150 mL Alkaline Phosphatase Reagent Kit
23-666-075 450 mL Alkaline Phosphatase Reagent Kit
23-666-076 625 mL Alkaline Phosphatase Reagent Kit
23-666-081 2 x 150 R2mL Alkaline Phosphatase R2 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 23-666-080 Supplier Pointe Scientific Supplier No. HA416R1

For quantitative determination of alkaline phosphatase in human serum

HA416-R1 and HA416-R2 are for Hitachi 747.

  • Methods: p-Npp, kinetic and for Hitachi 747
  • Linearity: 0 to 1000U/L
  • Wavelength: 405nm
  • Stability: 7 days at room temp. and 14 days at 2 to 8°C
  • Reaction Temp.: 3°C
  • Expected Values: Adults 35-123 U/L, Children have a higher normal value

Specifications

Certifications/Compliance CE Marked
Type Alkaline Phosphatase R1 Kit
Formulation R1 and R2 contains AMP Buffer pH 10.45, p-NPP ≤16mM, Magnesium Ions <1mM, activators and preservatives
For Use With (Application) Quantitative determination of Alkaline Phosphatase in Human Serum, in-vitro diagnostic use only
Quantity 5 x 500 R1mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.