Learn More
Description
HA416-R1 and HA416-R2 are for Hitachi 747.
- Methods: p-Npp, kinetic and for Hitachi 747
- Linearity: 0 to 1000U/L
- Wavelength: 405nm
- Stability: 7 days at room temp. and 14 days at 2 to 8°C
- Reaction Temp.: 3°C
- Expected Values: Adults 35-123 U/L, Children have a higher normal value
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Alkaline Phosphatase R2 Kit |
| Formulation | R1 and R2 contains AMP Buffer pH 10.45, p-NPP ≤16mM, Magnesium Ions <1mM, activators and preservatives |
| For Use With (Application) | Quantitative determination of Alkaline Phosphatase in Human Serum, in-vitro diagnostic use only |
| Quantity | 2 x 150 R2mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.