Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Glucose (Hexokinase) Liquid Reagents

Catalog No. 23666279 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120 mL
1 x 500 mL
1 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-279 1 x 1 L
23-666-278 1 x 120 mL
23-666-280 1 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-666-279 Supplier Pointe Scientific Supplier No. G75171L
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the in vitro quantitative measurement of glucose in serum.

  • Reagent requires Glucose Standard (23-666-277)/Chemistry Calibrator (23-666-189)

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Type Glucose (Hexokinase) Reagent Kit
Formulation Glucose reagent: Nicotinamide Adenine Dinucleotide 2mM/L, Adenosine Triphosphate 4mM/L, Magnesium 2mM/L, Hexokinase (Yeast) > 2000U/L, Glucose-6-Phosphate Sehydrogenase (microbial) >4000U/L, Stabilizers and preservatives
For Use With (Application) Quantitative measurement of Glucose in Serum
Quantity 1 x 1 L
Wavelength 340 nm
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.