Learn More
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | gamma-Glutamyl Transferase Reagent Kit |
| Formulation | Tris Buffer 89mM/L pH 8.1, Glycylglycine 126mM/L, GLUPA-C 3.3mM/L, Sodium Azide 0.095% |
| For Use With (Application) | Quantitative kinetic determination of gamma Glutamyl Transferase activity in Serum |
| Quantity | 3 x 120mL R1, 1 x 90mL R2 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.