Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific ISE 1N KCL Solution Reagents

Catalog No. 23666322 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
5 x 300 mL
1 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-322 5 x 300 mL
23-666-325 1 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-666-322 Supplier Pointe Scientific Supplier No. HI990KCL
This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Marked
Type 1N KCL Solution Reagent Kit
Formulation Potassium Chloride 1mol/L, Anti-microbial 0.01%
For Use With (Application) Quantitative determination of Sodium, Potassium and Chloride in Serum
Quantity 5 x 300 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.