Learn More
Description
Includes
Borate buffer: Sodium chloride 3.2mM; Sodium bicarbonate 1.3mM; Potassium phosphate 0.15mM; anti-microbial (0.05%)
- Method: Ion-Selective Electrode (ISE)
- Format: Liquid
ISE Diluent, ISE Internal Reference and ISE 1N KCl are for the quantitative determination of sodium, potassium, and chloride in serum by ion-selective electrode methods on Roche/Hitachi systems
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | ISE Internal Reference Solution |
| Formulation | ISE Internal Reference: Borate Buffer- Sodium Chloride 3.2mM/L, Sodium Bicarbonate 1.3mM/L, Potassium Phosphate 0.15mM/L, Anti-microbial Nonreactive Ingredient 0.05% |
| For Use With (Application) | Quantitative determination of Sodium, Potassium and Chloride in Serum |
| Quantity | 6 x 2 L |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.