Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific ISE Solution Reagents for Roche Cobas™ Analyzers

Catalog No. 23666449 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
60 mL
125 mL
250 mL
Formulation:
Ammonium Hydrogen Difluoride 0.6%
Aqueous solution containing: Na+ 100mM/L, K+ 7mM/L, Cl- 92mM/L and a preservative
Aqueous solution containing: Na+ 150mM/L, K+ 3mM/L, Cl 138mM/L and a preservative
KCl aqueous solution 3.5mM/L
Type:
ISE Solution Reagent
ISE Standard 1 Solution Reagent
ISE Standard 2 Solution Reagent
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation Type
23-666-449 250 mL Aqueous solution containing: Na+ 150mM/L, K+ 3mM/L, Cl 138mM/L and a preservative ISE Standard 1 Solution Reagent
23-666-448 60 mL KCl aqueous solution 3.5mM/L ISE Solution Reagent
23-666-447 125 mL Ammonium Hydrogen Difluoride 0.6% ISE Solution Reagent
23-666-450 60 mL Aqueous solution containing: Na+ 100mM/L, K+ 7mM/L, Cl- 92mM/L and a preservative ISE Standard 2 Solution Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-666-449 Supplier Pointe Scientific Supplier No. MIS601

Needed for quantification of sodium, potassium and chloride on Roche™ Cobas systems

  • Method: Ion-selective Electrode (ISE)
  • Format: Liquid

  • Reference Solution contains: KCl 3.5mM
  • Standard Solution 1 contains: Na+ 150mM; K+ 3mM; Cl- 138mM; Preservative
  • Standard Solution 2 contains: Na+ 100mM; K+ 7mM; Cl- 92mM; Preservative
  • Etcher Solution contains: Ammonium hydrogen difluoride 0.6%

For ISE applications on Roche Cobas Chemistry Systems

Specifications

Certifications/Compliance CE Marked
Type ISE Standard 1 Solution Reagent
Formulation Aqueous solution containing: Na+ 150mM/L, K+ 3mM/L, Cl 138mM/L and a preservative
For Use With (Application) To provide calibration points for the Sodium, Potassium and Chloride electrodes on the ISE module
Quantity 250 mL
Form Liquid

For in vitro diagnostic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.