Learn More
Pointe Scientific Lactate Dehydrogenase Reagents for Hitachi™ Analyzers
Shop All Pointe Scientific ProductsDescription
Includes
Reagent 1 and Reagent 2 when combined: NAD 7.5mM; L-Lactate 55mM; Buffer (pH 8.95); Non-reactive stabilizers; Sodium azide 0.1%
- Method: Modified Wacker/Tris, Kinetic
- Format: Liquid (ready to use)
- Linearity: 1000 U/L
- Expected Values: 50 to 166 U/L (Male, 30°C); 60 to 132 U/L (Female, 30°C)
Applications:
For the quantitative kinetic determination of lactate dehydrogenase activity in serum on Hitachi analyzers
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Lactate Dehydrogenase Reagent |
| Formulation | NAD 7.5mM, L-Lactate 55mM, Buffer pH 8.95, non-reactive Stabilizers and Sodium Azide 0.1% as preservative |
| For Use With (Application) | Quantitative kinetic determination of Lactate Dehydrogenase activity in Serum, in-vitro diagnostic use only |
| pH | 9 |
| Quantity | 4 x 50mL R1, 4 x 21mL R2 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.