Learn More
Description
Includes
4-Chlorophenol 3.5mM; ATP >0.5mM; Magnesium salt 10 mM; 4-Aminophenazone 0.3mM; Glycerol Kinase (microbial) >250U/L; Glycerol Phosphate Oxidase (microbial) >4500U/L; Peroxidase (horseradish) >2000U/L; Lipase (microbial) >200,000U/L; buffer (pH 7.3 ± 0.1); surfactants; stabilizers; preservatives, including sodium azide (0.01%)
Requires
Requires calibrator/standard
- Method: Enzymatic/GPO Trinder, Endpoint
- Format: Liquid (ready to use)
- Linearity: 1000mg/dL
- Expected Values: 44 to 148mg/dL
For the quantitative determination of triglycerides in serum or plasma on Hitachi analyzers
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Triglyceride Reagent Kit |
| For Use With (Application) | Quantitative determination of Triglyceride in Serum or Plasma, in-vitro diagnostic use only |
| pH | 7.3 |
| Quantity | 600 mL |
| Form | Liquid |
For in vitro diagnostic use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.