Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pokemon (LRF) Monoclonal Antibody (13E9), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7069414
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-130-22 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-130-22 Supplier Invitrogen™ Supplier No. 14330982
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: This monoclonal antibody reacts with human Leukemia/lymphoma-related factor (LRF). This 65-kDa protein is also known as POKEMON, FBI-1, and Zbtb7a. LRF is a member of the POK family of transcriptional repressors, and is expressed in hematopoietic stem cells, T and B lymphocytes, macrophages, and erythroid cells. This protein is also highly expressed in human non-Hodgkin's lymphoma tissues and diffuse large B-cell lymphomas, demonstrating its prominent role in oncogenic transformation. Studies have demonstrated that LRF is also involved in B cell lineage commitment and development by opposing Notch signaling. Moreover, LRF is critical for erythroid terminal differentiation in the bone marrow by inhibiting Bim-mediated apoptosis. Finally, LRF interacts with Bcl-6 in germinal center B cells. Applications Reported: This 13E9 antibody has been reported for use in immunoblotting (WB) and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This 13E9 antibody has been tested by western blot analysis of the A20 cell line and by immunohistochemical staining of formalin-fixed paraffin embedded human tonsil tissue. This can be used at 1-10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Plays a key role in the instruction of early lymphoid progenitors to develop into B lineage by repressing T-cell instructive Notch signals. Specifically represses the transcription of the CDKN2A gene. Efficiently abrogates E2F1-dependent CDKN2A transactivation/de-repression. Binds to the consensus sequence 5'-[GA][CA]GACCCCCCCCC-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pokemon (LRF)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 13E9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ZBTB7A
Gene Accession No. O95365
Gene Alias 9030619K07Rik; 9130006G12Rik; AI452336; cLRF; DKFZp547O146; factor binding IST protein 1; Factor that binds to inducer of short transcripts protein 1; FBI1; FBI-1; HIV-1 1st-binding protein 1; HIV-1 inducer of short transcripts binding protein; leukemia/lymohoma related factor cLRF; leukemia/lymphoma related factor; leukemia/lymphoma related factor cLRF; leukemia/lymphoma-related factor; LRF; lymphoma related factor; MGC99631; Oczf; Osteoclast-derived zinc finger protein; POK erythroid myeloid ontogenic factor; Pokemon; POZ and Krueppel erythroid myeloid ontogenic factor; TIP21; TTF-I-interacting peptide 21; Zbtb7; ZBTB7A; zi; zinc finger and BTB domain containing 7a; zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcripts binding protein; zinc finger and BTB domain-containing protein 7A; Zinc finger protein 857A; ZNF857A
Gene Symbols ZBTB7A
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51341
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.