Learn More
Pol kappa Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301975A

Description
The recommended shelf life for this product is 1 year from date of receipt.
External and internal DNA-damaging agents continually threaten the integrity of genetic material in cells. Although a variety of repair mechanisms exist to remove the resulting lesions, some lesions escape repair and block the replication machinery. Members of the Y family of DNA polymerases, such as POLK, permit the continuity of the replication fork by allowing replication through such DNA lesions. Each Y family polymerase has a unique DNA-damage bypass and fidelity profile. POLK is specialized for the extension step of lesion bypass .Specifications
Pol kappa | |
Polyclonal | |
Unconjugated | |
POLK | |
Polymerase (DNA directed) kappa, DINB1, DINP, POLQ | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51426 | |
4° C | |
Liquid |
Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9UBT6 | |
POLK | |
Between 675 and 725 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.