Learn More
Pol Lambda Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | Pol Lambda |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1902
![]() |
Bethyl Laboratories, Inc
IHC00325T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1901
![]() |
Bethyl Laboratories, Inc
IHC00325 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Gorilla, Chimpanzee
This gene encodes a DNA polymerase. DNA polymerases catalyze DNA-template-directed extension of the 3'-end of a DNA strand. This particular polymerase, which is a member of the X family of DNA polymerases, likely plays a role in non-homologous end joining and other DNA repair processes. Alternatively spliced transcript variants have been described.Specifications
Pol Lambda | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
POLL; BETA-N; POL-KAPPA; polymerase (DNA directed), lambda; polymerase (DNA-directed), lambda; DNA polymerase kappa (DNA polymerase beta-N); RP11-529I10.1; BETAN; FLJ46002; POLKAPPA; DNA polymerase beta-2; DNA polymerase kappa (DNA polymerase beta-N); DNA polymerase lamda2; DNA-directed DNA polymerase lambda; OTTHUMP00000020323; pol beta2; pol lambda | |
Between 525 and 575 | |
Primary | |
4° C | |
POLL |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9QXE2, Q9UGP5 | |
27343, 56626 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.