Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pol Lambda Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560883
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0883 Supplier Bethyl Laboratories, Inc Supplier No. A301640A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes a DNA polymerase. DNA polymerases catalyze DNA-template-directed extension of the 3′-end of a DNA strand. This particular polymerase, which is a member of the X family of DNA polymerases, likely plays a role in non-homologous end joining and other DNA repair processes. Alternatively spliced transcript variants have been described.

Specifications

Antigen Pol Lambda
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene POLL
Gene Accession No. Q9UGP5
Gene Alias POLL; BETA-N; POL-KAPPA; polymerase (DNA directed), lambda; polymerase (DNA-directed), lambda; DNA polymerase kappa (DNA polymerase beta-N); RP11-529I10.1; BETAN; FLJ46002; POLKAPPA; DNA polymerase beta-2; DNA polymerase kappa (DNA polymerase beta-N); DNA polymerase lamda2; DNA-directed DNA polymerase lambda; OTTHUMP00000020323; pol beta2; pol lambda
Gene Symbols POLL
Host Species Rabbit
Immunogen Between 150 and 200
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27343
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.