Learn More
Pol Lambda Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301641AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat
This gene encodes a DNA polymerase. DNA polymerases catalyze DNA-template-directed extension of the 3′-end of a DNA strand. This particular polymerase, which is a member of the X family of DNA polymerases, likely plays a role in non-homologous end joining and other DNA repair processes. Alternatively spliced transcript variants have been described.Specifications
| Pol Lambda | |
| Polyclonal | |
| Unconjugated | |
| POLL | |
| POLL; BETA-N; POL-KAPPA; polymerase (DNA directed), lambda; polymerase (DNA-directed), lambda; DNA polymerase kappa (DNA polymerase beta-N); RP11-529I10.1; BETAN; FLJ46002; POLKAPPA; DNA polymerase beta-2; DNA polymerase kappa (DNA polymerase beta-N); DNA polymerase lamda2; DNA-directed DNA polymerase lambda; OTTHUMP00000020323; pol beta2; pol lambda | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27343, 56626 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9QXE2, Q9UGP5 | |
| Poll | |
| Between 525 and 575 | |
| 10 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.