Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PolA1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | PolA1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-3637
![]() |
Bethyl Laboratories, Inc
A302850AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
DNA replication, recombination and repair, all of which are necessary for genomic stability, require the presence of exonucleases. In DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair they function to excise damaged DNA fragments and correct recombinational mismatches. These exonucleases include the family of DNA polymerases. DNA pol alpha, beta, delta and epsilon are involved in DNA replication and repair. DNA pol delta and DNA pol epsilon are multisubunit enzymes, with DNA pol delta consisting of two subunits: p125, which interacts with the sliding DNA clamp protein PCNA; and p50. The nuclear-encoded DNA pol gamma is the only DNA polymerase required for the replication of the mitochondrial DNA. DNA pol zeta is ubiquitously expressed in various tissues and mediates the cellular mechanism of damage-induced mutagenesis. DNA pol theta is a DNA polymerase-helicase that binds ATP and is involved in the repair of interstrand crosslinks.Specifications
PolA1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
DNA polymerase alpha catalytic subunit, DNA polymerase alpha catalytic subunit p180, POLA1, POLA, DNA pol alpha | |
Between 775 and 825 | |
Primary | |
4° C | |
POLA1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P09884, P33609 | |
18968, 5422 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title