Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLA2 Antibody (OTI3F2), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273514AF405
Description
POLA2 Monoclonal specifically detects POLA2 in Human samples. It is validated for Western Blot.Specifications
POLA2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
POLA2 | |
Recombinant protein fragment corresponding to amino acids 1-284 of human POLA2 (NP_002680) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI3F2 | |
Western Blot | |
DNA polymerase alpha 70 kDa subunit, DNA polymerase alpha subunit B, FLJ21662, FLJ37250, polymerase (DNA directed), alpha 2 (70kD subunit), polymerase (DNA-directed), alpha (70kD) | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
23649.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction