Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558015 Supplier Invitrogen™ Supplier No. PA558015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IMEGINTTGR KLVATKLYEG VPLPFYQPTE EDADFEQSMV LVACGPYTTS DSITYDPLLD LIAVINHDRP DVCILFGPFL DAKHEQVENC L Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 90%.

The protein encoded by this gene is a zinc finger protein that may act as a transcription factor. The encoded protein may be part of a BHC histone deacetylase complex. Translocation of this gene with the fibroblast growth factor receptor-1 gene (FGFR1) results in a fusion gene, which may be a cause of stem cell leukemia lymphoma syndrome (SCLL). Several transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLA2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLA2
Gene Accession No. Q14181
Gene Alias AI573378; DNA polymerase alpha 2, 68 kDa; DNA polymerase alpha 2, accessory subunit; DNA polymerase alpha 70 kDa subunit; DNA polymerase alpha subunit B; DNA polymerase alpha subunit II; DNA polymerase subunit II; DNA-directed DNA polymerase alpha 2; Ese1L; Pola2; polymerase (DNA directed), alpha 2; polymerase (DNA directed), alpha 2 (70kD subunit); polymerase (DNA directed), alpha 2, accessory subunit; polymerase (DNA) alpha 2, accessory subunit; polymerase (DNA-directed), alpha (70kD)
Gene Symbols POLA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLA2. Recombinant protein control fragment (Product #RP-97265).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.