Learn More
POLA2 Polyclonal Antibody

Description
POLA2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | POLA2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | POLA2 |
| Gene Accession No. | P33611 |
| Gene Alias | AI573378; DNA polymerase alpha 2, 68 kDa; DNA polymerase alpha 2, accessory subunit; DNA polymerase alpha 70 kDa subunit; DNA polymerase alpha subunit B; DNA polymerase alpha subunit II; DNA polymerase subunit II; DNA-directed DNA polymerase alpha 2; Ese1L; Pola2; polymerase (DNA directed), alpha 2; polymerase (DNA directed), alpha 2 (70kD subunit); polymerase (DNA directed), alpha 2, accessory subunit; polymerase (DNA) alpha 2, accessory subunit; polymerase (DNA-directed), alpha (70kD) |
| Gene Symbols | POLA2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.