Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argos Technologies™ Cardboard Cryogenic Storage Box Dividers, 64-place

Catalog No. 03391510 Shop All Oakton Products
Change view
Click to view available options
Holds:
64 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
03-391-510 64 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-391-510 Supplier Argos Technologies™ Supplier No. 0675583
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Dividers only. (Boxes are sold separately.)

Create customized storage box configurations

  • Snug-fitting lid prevents accidental separation if dropped
  • Use in mechanical freezers or liquid nitrogen tanks to –196°C
  • Constructed of sturdy cardboard with moisture-repellent coating
  • Designed with small holes on the bottom to allow for drainage

These economical cryo/freezer storage box dividers offer excellent low-temperature storage. Moisture-repellent coating protects them from effects of liquid nitrogen or condensation. Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL. Boxes can be ordered with dividers for a complete box configuration or without dividers to customize your storage needs. The wide range of dividers can hold from 16 to 196 tubes with diameters from 8 to 33.5 mm.

Specifications

Description 64-place cell divider
Color White
Material Fiberboard
Shape Square
Type Cell Dividers for Standard Tubes
For Use With (Application) Sample Storage for Laboratories in Universities and for Performing Artificial Insemination, Blood Bank, and Stem Cell/Bone Marrow Procedures in Institutions
Holds 64 Tubes
Quantity 12 Pack
Dimensions (L x W x H) 4.8 x 4.8 x 1.3 in.
Diameter (Metric) Well 14.6 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.