Learn More
Argos Technologies™ Fiberboard Cryogenic Storage Boxes and Dividers
Catalog No. 03391515 Shop All Oakton Products
Change view
Click to view available options
Holds:
81 Tubes
Catalog No. Holds
03-391-515 81 Tubes
1 options
Catalog No. 03-391-515 Supplier Argos Technologies™ Supplier No. 0675572
Add to Cart
Add to Cart
Create customized storage box configurations
  • Snug-fitting lid prevents accidental separation if dropped
  • Use in mechanical freezers or liquid nitrogen tanks to –196°C
  • Constructed of sturdy cardboard with moisture-repellent coating
  • Designed with small holes on the bottom to allow for drainage
These economical cryo/freezer storage boxes offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation.
Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL. Boxes can be ordered with dividers for a complete box configuration or without dividers to customize your storage needs. The wide range of dividers can hold from 16 to 196 tubes with diameters from 8 to 33.5 mm. Order additional and replacement cell dividers separately.
Specifications
Description 81-place cell divider
Color White
Material Fiberboard
Shape Square
Type Freezer Box
For Use With (Application) Blood Bank, Sample Storage for Laboratories in Universities and for Performing Artificial Insemination, Stem Cell/Bone Marrow Procedures
Holds 81 Tubes
Dimensions (L x W x H) 5.2 x 5.2 x 2 in.