Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argos Technologies™ PolarSafe™ Cryogenic Vials

Catalog No. p-7110994
Click to view available options
Volume (Metric):
1 mL
2 mL
3 mL
4 mL
5 mL
Base Style:
Round
Self-standing
Height (Metric):
41 mm
45 mm
46 mm
69 mm
74 mm
88 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Innovactive screw cap eliminates need for silicone o-rings

  • Designed to withstand temperature from -196 to 121°C
  • Printed barcode and white writing surface aid in specimen identification
  • Certified at 95 kPa to provide a leakproof seal

Cryogenic storage vials feature a co-molded screw cap that combines polyethylene (PE) with a thermoplastic elastomer (TPE), eliminating silicone O-rings for a safer sample storage environment. Cap design provides a secure seal on the inside of the vial, makes gripping the outside of the cap easier, and works well with automatic decapper machines. Caps are offered with external or internal threaded configurations. Multicolored cap inserts (not included) are available to aid in visual identification.

Vials are available with round or self-standing skirted bottoms. A special locking system on the bottom of each skirted vial interlocks to a cryogenic rack (not included) for easy one-handed manipulation.

The vials are manufactured in a ISO Class 7 cleanroom in accordance with FDA, GMP, and ISO quality standards. Sterilized by gamma irradiation, they are packaged in resealable bags of 50. They are also certified to be DNase, RNase, pyrogen, ATP and human DNA free.

Do not use vials for storage in the liquid phaes of liquid nitrogen: this may cause entrapment of liquified nitrogen inside the vial and lead to pressure build-up,resulting in possible explosion or biohazard release.

Specifications

Color Clear
Diameter (Metric) Outer 12.5 mm
Material Polyethylene
Pre-slit No
Sterility Sterile
Volume (Metric) 1 mL
Height (English) 1.6 in.
Temperature Range (Metric) -196°C to +121°C
Thread Style External
Base Style Self-standing
RNase Free Yes
Dnase Free Yes
Barcode Serial/consecutive Barcode128 printed on each tube
Closure Material Polyethylene, Thermoplastic Elastomer
Autoclavable Autoclavable
Closure Type Screw Cap
Height (Metric) 41 mm
Quantity 50/Cs.
Certifications/Compliance FDA, GMP and ISO quality standards (ISO13485:2004, ISO14644 and 14698)
Type Cryogenic Vial
Preservative None
Marking Spot Yes
For Use With (Equipment) Cryogenic Storage
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.