Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLB Recombinant Rabbit Monoclonal Antibody (6M8I5)
GREENER_CHOICE

Catalog No. PIMA535359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535359 Supplier Invitrogen™ Supplier No. MA535359
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MGVCQLPSKN DEKEYPHRRI DIRLIPKDQY YCGVLYFTGS DIFNKNMRAH ALEKGFTINE YTIRPLGVTG VAGEPLPVDS EKDIFDYIQW KYREPKDRSE.

DNA polymerase beta comprises an amino-terminal 8- kDa domain and a carboxy-terminal 31- kDa domain. The N-terminal ssDNA binding domain has a deoxyribose phosphodiesterase activity while the C-terminal domain has a nucleotidyltransferase activity. Mammalian DNA polymerase beta, a DNA repair polymerase, is constitutively expressed in cultured cells, but treatment of cells with the DNA-alkylating agents such as N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) or methyl methanesulfonate up-regulates beta-pol level. DNA polymerase beta fills single nucleotide gaps in DNA produced by the base excision repair pathway of mammalian cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 6M8I5
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POLB
Gene Accession No. P06746, P06766, Q8K409
Gene Alias A430088C08Rik; DNA pol beta; DNA polymerase beta; DNA polymerase beta subunit; DNA-directed DNA polymerase beta; hypothetical protein LOC614688; LOW QUALITY PROTEIN: DNA polymerase beta; MGC125976; POL B; Pol beta; POLB; polb.S; polymerase (DNA directed), beta; polymerase (DNA directed), beta S homeolog; polymerase (DNA) beta; polymerase B; polymerase-beta; wu:fa97a05; XELAEV_18020165mg; zgc:109924
Gene Symbols POLB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 236-335 of human DNA Polymerase beta (P06746).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18970, 29240, 5423
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.