Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583823
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583823 Supplier Invitrogen™ Supplier No. PA583823
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YALRLKEKAT QCQLEADVLW SDVVSHPPEG PWQRIAPLRV LSFDIECAGR KGIFPEPERD PVIQICSLGL RWGEPEPFLR LALTLRPCAP ILGAKVQSYE KEEDLLQAWS TFIRIMDPD Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 88%.

This gene encodes the 125-kDa catalytic subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3' to 5' exonuclease activity and plays a critical role in DNA replication and repair. Alternatively spliced transcript variants have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLD1
Gene Accession No. P28340
Gene Alias 125kDa; CDC2; CDC2 homolog; CRCS10; DAN polymerase delta 1, catalytic subunit; DNA polymerase delta 1, catalytic domain; DNA polymerase delta catalytic subunit; DNA polymerase delta, catalytic subunit; DNA polymerase subunit delta p125; DNA-directed DNA polymerase delta 1; MDPL; POL D; POLD; POLD1; polymerase (DNA directed), delta 1, catalytic subunit; polymerase (DNA directed), delta 1, catalytic subunit 125kDa; polymerase (DNA) delta 1, catalytic subunit; Polymerase D
Gene Symbols POLD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLD1. Recombinant protein control fragment (Product #RP-89465).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5424
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.