Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POLD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555401 Supplier Invitrogen™ Supplier No. PA555401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRSKYIHPDD ELVLEDELQR IKLKGTIDVS KLVTGTVLAV FGSVRDDGKF LVEDYCFADL APQKPAPPLD TDRFVLLVSG LGLGGGGGES LLGTQLLVDV VTGQLGDEGE QC Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.

The function of the small subunit is not yet clear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLD2
Gene Accession No. P49005
Gene Alias 50kDa; DAN polymerase delta 2, accessory subunit; DNA polymerase delta subunit 2; DNA polymerase delta subunit p50; DNA-directed DNA polymerase delta 2; p50; po1D2; Pol delta B subunit (p50); POLD2; polymerase (DNA directed), delta 2, accessory subunit; polymerase (DNA directed), delta 2, regulatory subunit; polymerase (DNA directed), delta 2, regulatory subunit 50kDa; polymerase (DNA) delta 2, accessory subunit
Gene Symbols POLD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLD2. Recombinant protein control fragment (Product #RP-92687).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5425
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.