Learn More
PolD2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304322A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The function of the small subunit is not yet clear.Specifications
PolD2 | |
Polyclonal | |
Unconjugated | |
POLD2 | |
POLD2;DNA polymerase delta subunit 2; DNA polymerase delta subunit 2; DNA polymerase delta subunit p50 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5425 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P49005 | |
POLD2 | |
Between 419 and 469 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.