Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLD4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | POLD4 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5259
![]() ![]() |
Proteintech Group Inc
262091AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes the smallest subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3′ to 5′ exonuclease activity and plays a critical role in DNA replication and repair. The encoded protein enhances the activity of DNA polymerase delta and plays a role in fork repair and stabilization through interactions with the DNA helicase Bloom syndrome protein. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
POLD4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9HCU8 | |
57804 | |
POLD4 Fusion Protein Ag23924 | |
Primary | |
-20°C | |
POLD4 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DNA polymerase delta subunit 4, p12, POLD4, POLDS | |
POLD4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title