Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLDIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214147
Description
POLDIP1 Polyclonal specifically detects POLDIP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
POLDIP1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
BACURD1, BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 1, BTB/POZ domain-containing protein KCTD13, hBACURD1, PDIP1FKSG86, POLDIP1TNFAIP1-like protein, Polymerase delta-interacting protein 1, potassium channel tetramerisation domain containing 13 | |
Rabbit | |
Affinity Purified | |
RUO | |
253980 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
KCTD13 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: RGEDEENREHRVRRIHVRRHITHDERPHGQQIVFKD | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction