Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POLDIP1 Antibody (OTI3B12), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273515AF488
Description
POLDIP1 Monoclonal specifically detects POLDIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
POLDIP1 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KCTD13 | |
Full length recombinant protein of human POLDIP1 (NP_849194) produced in E.coli. | |
0.1 mL | |
253980.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3B12 | |
Western Blot, Immunohistochemistry | |
BACURD1, BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 1, BTB/POZ domain-containing protein KCTD13, hBACURD1, PDIP1FKSG86, POLDIP1TNFAIP1-like protein, Polymerase delta-interacting protein 1, potassium channel tetramerisation domain containing 13 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction